Learn More
Enzo Life Sciences Human Genomic DNA, Female (100µg)
SDP
Catalog No. 502013751
Encompass_Preferred
Human Genomic DNA, Female, is comprised of a pool of ≥ 8 normal donors. It is intact DNA extracted from freshly harvested whole blood of healthy females. DNA is treated with DNase free-RNase to remove residual contaminant RNA. Human genomic DNA can be used for a variety of molecular applications including PCR, southern blotting, and DNA library construction. It can also be used as normal reference DNA for comparative genomic hybridizations (CGH). Alternative name: gDNA. Concentration: 150 - 250 µg/mL in 10 mM Tris-HCl, pH 8.0, containing 1 mM EDTA. Formulation: Liquid. Long Term Storage: -20°C.
Catalog No. 50-201-3751 Supplier Enzo Life Sciences Supplier No. ENZGEN1180100
Human Genomic DNA, Female, is comprised of a pool of ≥ 8 normal donors. It is intact DNA extracted from freshly harvested whole blood of healthy females. DNA is treated with DNase free-RNase to remove residual contaminant RNA. Human genomic DNA can be used for a variety of molecular applications including PCR, southern blotting, and DNA library construction. It can also be used as normal reference DNA for comparative genomic hybridizations (CGH). Alternative name: gDNA. Concentration: 150 - 250 µg/mL in 10 mM Tris-HCl, pH 8.0, containing 1 mM EDTA. Formulation: Liquid. Long Term Storage: -20°C.