Learn More
Sino Biological Recombinant Human GFAP Protein (His Tag)
Catalog No. 502993537
Encompass_Preferred
A DNA sequence encoding the Human GFAP (P14136) (Met1-Met432) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-299-3537 Supplier Sino Biological Supplier No. 12167H07E100
A DNA sequence encoding the Human GFAP (P14136) (Met1-Met432) was expressed, with a polyhistidine tag at the N-terminus.