Learn More
ACROBiosystems Human GFAP, His Tag (GFP-H5143) is expressed from E.coli cells. It contains AA Met 1 - Met 432 (Accession # P14136-1).
Supplier:  ACROBiosystems GFPH514325UG
Encompass_Preferred
GFAP (Glial fibrillary acidic protein) is a monomeric intermediate filament protein fuond in mature astrocytes and other glial cells but is not found outside the CNS. Increased GFAP immunoreactivity (or astrocytic activation) is usually viewed as an index of gliosis or a relatively slow-developing correlate of neural damage.
Catalog No. 50-225-3883