Learn More
Bon Opus Biosciences Recombinant Human GFAP protein (N-His) - Source: E. coli
SDP
Catalog No. NC2733755
Encompass_Preferred
Recombinant Human Glial Fibrillary Acidic Protein is produced by our E.coli expression system and the target gene encoding Leu292-Met432 is expressed with a 6His tag at the N-terminus. Accession#P14136. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4
Catalog No. NC2733755 Supplier Bon Opus Biosciences Supplier No. BP27250UG
Recombinant Human Glial Fibrillary Acidic Protein is produced by our E.coli expression system and the target gene encoding Leu292-Met432 is expressed with a 6His tag at the N-terminus. Accession#P14136. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4