Learn More
Sino Biological Anti-GFPT1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11071RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human GFAT (rh GFAT; Catalog#11071-HNAE; AAA58502.1; Gln 332-Glu 699). GFAT specific IgG was purified by human GFAT affinity chromatography.
Catalog No. 50-240-9674