Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human GFR alpha-3 ELISA Kit

Catalog No. EH218RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH218RBX5 5 x 96 Tests
EH218RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH218RBX5 Supplier Invitrogen™ Supplier No. EH218RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Human GFR alpha-3 quantitates human GFR alpha-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GFR alpha-3.

GFRa-3 binds persephin, thus, persephin, GFRa-3, and Ret PTK form a complex to transduce persephin signal and to mediate persephin function.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O60609
Assay Range 16.38-4000 pg/mL
Assay Sensitivity 16 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias GDNF family receptor alpha 3; GDNF family receptor alpha-3; GDNF receptor alpha-3; GDNF-family receptor alpha 3; GDNFR3; GDNFR-alpha-3; GFR alpha-3; GFRA3; GFRalpha3; GFR-alpha-3; glial cell line derived neurotrophic factor family receptor alpha 3; glial cell line-derived neurotrophic factor receptor alpha-3; GPI-linked receptor; UNQ339/PRO538/PRO3664; Y15110GDNF family receptor alpha 3; GDNF family receptor alpha-3; GDNF receptor alpha-3; GDNF-family receptor alpha 3; GDNFR3; GDNFR-alpha-3; GFR alpha-3; GFRA3; GFRalpha3; GFR-alpha-3; glial cell line derived neurotrophic factor family receptor alpha 3; glial cell line-derived neurotrophic factor receptor alpha-3; GPI-linked receptor; UNQ339/PRO538/PRO3664; Y15110;
Gene ID (Entrez) 2676
Gene Symbol Gfra3
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target GFR alpha-3
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.