Learn More
Sino Biological Recombinant Human GFR Alpha-1/GFRA1 Protein (hFc Tag)
Supplier:  Sino Biological 10330H02H100
Encompass_Preferred
A DNA sequence encoding the human GFRA1 (NP_001138925.1) (Met1-Ser424) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-5375