Learn More
Bon Opus Biosciences Recombinant Human GFRAL-D3 (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP110450UG
Encompass_Preferred
Recombinant Human GFRAL-D3 (C-hFc) is produced by Human Cells. The target gene encoding Cys210-Glu351 is expressed with a C-hFc tag. Accession# Q6UXV0-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1104
Catalog No. 50-217-2701