Learn More
Sino Biological Recombinant Human GFRAL Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 27245H02H1
Encompass_Preferred
A DNA sequence encoding the Human GFRAL (NP_997293.2) (Ser19-Glu351) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-3837