Learn More
Sino Biological Recombinant Human GFRAL Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 27245H08H1
Encompass_Preferred
A DNA sequence encoding the Human GFRAL (NP_997293.2) (Ser19-Glu351) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3835