Learn More
Sino Biological Anti-GGT1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100416T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human GGT1, and purified by antigen affinity chromatography.
Catalog No. 50-241-2256