Learn More
Sino Biological Recombinant Anti-Growth Hormone Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 16122R177100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Growth Hormone / GH1 (rh Growth Hormone / GH1; Catalog#16122-H07E; P01241; Phe27-Phe217).
Catalog No. 50-255-4038