Learn More
Sino Biological Anti-Growth Hormone Antibody, Mouse Monoclonal
Supplier:  Sino Biological 16122MM09100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Growth Hormone (rh Growth Hormone; Catalog#16122-H07E; P01241; Phe27-Phe217). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4042