Learn More
Raybiotech Inc Human Growth Hormone R ELISA Kit
Supplier:  Raybiotech Inc ELHGHR1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.4 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes human Growth Hormone R. Other species not yet determined.
Catalog No. 50-194-9312