Learn More
Invitrogen™ Human GHR ELISA Kit
Description
Human GHR quantitates human GHR in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GHR.
Specifications
Specifications
| Accession Number | P10912 |
| Assay Range | 0.4-100 ng/mL |
| Assay Sensitivity | 0.4 ng/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | GH receptor; GH-binding protein; GHBP; GHIP; GHR; GHR/BP; growth hormone binding protein; growth hormone receptor; growth hormone receptor precursor; Growth hormone receptor precursor (GH receptor) (GH binding protein) (GHBP) (Serum binding protein); growth hormone receptor precursor splice variant D56; growth hormone receptor variant d5-6; growth hormone receptor/binding protein; Growth hormone-binding protein; MGC124963; MGC156665; serum binding protein; Serum-binding protein; Somatotropin receptor; unnamed protein productGH receptor; GH-binding protein; GHBP; GHIP; GHR; GHR/BP; growth hormone binding protein; growth hormone receptor; growth hormone receptor precursor; Growth hormone receptor precursor (GH receptor) (GH binding protein) (GHBP) (Serum binding protein); growth hormone receptor precursor splice variant D56; growth hormone receptor variant d5-6; growth hormone receptor/binding protein; Growth hormone-binding protein; MGC124963; MGC156665; serum binding protein; Serum-binding protein; Somatotropin receptor; unnamed protein product; |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.