Learn More
Sino Biological Recombinant Anti-Intrinsic Factor/CBLIF Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 13544R01620
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Gastric intrinsic factor / GIF (rh Gastric intrinsic factor / GIF; Catalog#13544-H08H; P27352-1; Met1-Tyr417).
Catalog No. 50-241-2410