Learn More
Sino Biological Anti-Intrinsic Factor/CBLIF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13544RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human GIF / Gastric intrinsic factor (rh GIF / Gastric intrinsic factor; Catalog#13544-H08H; P27352-1; Met1-Tyr417). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2591