Learn More
Biomatik Corporation Human Gigaxonin (GAN) ELISA Kit
Supplier:  Biomatik Corporation EKU04371
Encompass_Preferred
Human Gigaxonin (GAN) ELISA Kit for detecting GAN in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H2C0. Gene ID: 8139. Target Synonyms: GAN1; KLHL16; Giant Axonal Neuropathy; Kelch-like protein 16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7028