Learn More
Medchemexpress LLC human GIP(3-30), amide | 1884226-05-4 | 99.8% | 3297.69 | 10 MG
SDP
Catalog No. 5000424584
Encompass_Preferred
Human GIP(3-30), amide is a high-affinity antagonist of the human GIP receptor in vitro, with potential anti-obesity and anti-diabetic effects. It competitively binds to the GIP receptor, inhibiting GIP-mediated cAMP accumulation, β-arrestin recruitment, and receptor internalization. It also inhibits GIP-mediated insulin secretion and signal transduction in adipocytes.
  • High-affinity antagonist of human GIP receptor.
  • Potential anti-obesity and anti-diabetic effects.
  • Inhibits GIP-mediated cAMP accumulation and insulin secretion.
  • Purity of 99.8%.
  • Powder storage: -80°C for 2 years or -20°C for 1 year.
  • For research use only.
Catalog No. 50-004-24584 Supplier Medchemexpress LLC Supplier No. HYP1013810MG
Human GIP(3-30), amide is a high-affinity antagonist of the human GIP receptor in vitro, with potential anti-obesity and anti-diabetic effects. It competitively binds to the GIP receptor, inhibiting GIP-mediated cAMP accumulation, β-arrestin recruitment, and receptor internalization. It also inhibits GIP-mediated insulin secretion and signal transduction in adipocytes.
  • High-affinity antagonist of human GIP receptor.
  • Potential anti-obesity and anti-diabetic effects.
  • Inhibits GIP-mediated cAMP accumulation and insulin secretion.
  • Purity of 99.8%.
  • Powder storage: -80°C for 2 years or -20°C for 1 year.
  • For research use only.