Learn More
Medchemexpress LLC Human GIP(3-30), amide TFA | 1884226-05-4 | 95.6% | 3297.69 | 5 MG
SDP
Catalog No. 5000424421
Encompass_Preferred
Human GIP(3-30), amide TFA is the TFA salt form of human GIP(3-30), amide. It is a high affinity antagonist of the human GIP receptor in vitro and has potential anti-obesity and anti-diabetic effects.
  • High affinity antagonist of the human GIP receptor in vitro.
  • Potential anti-obesity and anti-diabetic effects.
  • Competitively binds to GIP receptor (GIPR), blocking GIP-mediated cAMP accumulation (Ki=16.8 nM), β-arrestin recruitment (Ki for β-arrestin-1/2 recruitment is 10.6 nM and 10.2 nM) and receptor internalization.
  • Inhibits GIP mediated insulin secretion and signal transduction in adipocytes.
Catalog No. 50-004-24421 Supplier Medchemexpress LLC Supplier No. HYP10138A5MG
Human GIP(3-30), amide TFA is the TFA salt form of human GIP(3-30), amide. It is a high affinity antagonist of the human GIP receptor in vitro and has potential anti-obesity and anti-diabetic effects.
  • High affinity antagonist of the human GIP receptor in vitro.
  • Potential anti-obesity and anti-diabetic effects.
  • Competitively binds to GIP receptor (GIPR), blocking GIP-mediated cAMP accumulation (Ki=16.8 nM), β-arrestin recruitment (Ki for β-arrestin-1/2 recruitment is 10.6 nM and 10.2 nM) and receptor internalization.
  • Inhibits GIP mediated insulin secretion and signal transduction in adipocytes.