Learn More
Medchemexpress LLC Human GIP(3-30), amide TFA | 95.6% | 3297.69 (free base) | 10 MG
SDP
Catalog No. 5000423447
Encompass_Preferred
Human GIP(3-30), amide TFA is the TFA salt form of human GIP(3-30), amide (HY-P10138). It is a high affinity antagonist of the human GIP receptor in vitro. Human GIP(3-30), amide TFA has potential anti-obesity and anti-diabetic effects.
  • High affinity antagonist of the human GIP receptor in vitro.
  • Potential anti-obesity effects.
  • Potential anti-diabetic effects.
  • Competitively binds to GIP receptor (GIPR), blocking GIP-mediated cAMP accumulation (Ki=16.8 nM), β-arrestin recruitment (Ki for β-arrestin-1/2 recruitment is 10.6 nM and 10.2 nM) and receptor internalization.
  • Inhibits GIP mediated insulin secretion and signal transduction in adipocytes.
Catalog No. 50-004-23447 Supplier Medchemexpress LLC Supplier No. HYP10138A10MG
Human GIP(3-30), amide TFA is the TFA salt form of human GIP(3-30), amide (HY-P10138). It is a high affinity antagonist of the human GIP receptor in vitro. Human GIP(3-30), amide TFA has potential anti-obesity and anti-diabetic effects.
  • High affinity antagonist of the human GIP receptor in vitro.
  • Potential anti-obesity effects.
  • Potential anti-diabetic effects.
  • Competitively binds to GIP receptor (GIPR), blocking GIP-mediated cAMP accumulation (Ki=16.8 nM), β-arrestin recruitment (Ki for β-arrestin-1/2 recruitment is 10.6 nM and 10.2 nM) and receptor internalization.
  • Inhibits GIP mediated insulin secretion and signal transduction in adipocytes.