Learn More
IBL America Gastric Inhibitory Polypeptide (GIP), Total (Human) ELISA
Supplier:  IBL America 27203
Encompass_Preferred
Solid phase sandwich ELISA for the determination of human total GIP in EDTA-plasma and cell culture supernatant. For research use only, not for use in diagnostic procedures.
Catalog No. 50-212-0320