Learn More
ACROBiosystems Human GIPR 200ug
Supplier:  ACROBiosystems GIRH82E3200UG
Encompass_Preferred
Biotinylated Human GIPR Protein HisAvitag (GIR-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Gln 138 (Accession # P48546-1).
Catalog No. 50-253-5021