Learn More
ACROBiosystems Human GIPR 25ug
Catalog No. 502535020
Encompass_Preferred
Biotinylated Human GIPR Protein HisAvitag (GIR-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Gln 138 (Accession # P48546-1).
Catalog No. 50-253-5020 Supplier ACROBiosystems Supplier No. GIRH82E325UG
Biotinylated Human GIPR Protein HisAvitag (GIR-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Gln 138 (Accession # P48546-1).