Learn More
Sino Biological Recombinant Human GIPR Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 18774H56HB100
Encompass_Preferred
A DNA sequence encoding the Human GIPR (NP_000155.1) (Gly26-Gln138) was expressed with a N-terminal polyhistidine tag followed by an AVI tag.The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-3990