Learn More
BIOHIPPO Human GKN2 Microsample
SDP
Catalog No. 5000314384
Encompass
Human GKN2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GKN2 in Tissue homogenates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 3 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GKN2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GKN2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GKN2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14384 Supplier BIOHIPPO Supplier No. BHE15211668
Human GKN2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GKN2 in Tissue homogenates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 3 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GKN2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GKN2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GKN2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color