Learn More
Sino Biological Human GLA/alpha-Galactosidase A Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000786750
Encompass_Preferred
The Human GLA/alpha-Galactosidase A ELISA Pair Set is for the quantitative determination of Human GLA/alpha-Galactosidase A.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human GLA/alpha-Galactosidase A coated on a 96-well plate. Standards and samples are added to the wells, and any Human GLA/alpha-Galactosidase A present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human GLA/alpha-Galactosidase A monoclonal antibody is then added, producing an antibody-antigen-antibody ""sandwich"". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human GLA/alpha-Galactosidase A present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86750 Supplier Sino Biological Supplier No. SEKA120785
The Human GLA/alpha-Galactosidase A ELISA Pair Set is for the quantitative determination of Human GLA/alpha-Galactosidase A.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human GLA/alpha-Galactosidase A coated on a 96-well plate. Standards and samples are added to the wells, and any Human GLA/alpha-Galactosidase A present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human GLA/alpha-Galactosidase A monoclonal antibody is then added, producing an antibody-antigen-antibody ""sandwich"". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human GLA/alpha-Galactosidase A present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.