Learn More
Sino Biological Human GLA ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502370892
Encompass_Preferred
Each vial contains 455 ng of recombinant Human alpha-Galactosidase A / GLA. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 13000 pg/mL is recommended.
Catalog No. 50-237-0892 Supplier Sino Biological Supplier No. SEK120785
Each vial contains 455 ng of recombinant Human alpha-Galactosidase A / GLA. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 13000 pg/mL is recommended.