Learn More
Sino Biological Anti-GLA/alpha-Galactosidase A Antibody, Mouse Monoclonal
Supplier:  Sino Biological 12078MM0520
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human alpha-Galactosidase A (rh alpha-Galactosidase A; Catalog#12078-H08H; NP_000160.1; Met1-Leu429). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-9566