Learn More
Sino Biological Anti-GLA/alpha-Galactosidase A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12078T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human alpha-Galactosidase A / GLA (rh alpha-Galactosidase A / GLA; Catalog#12078-H08H; NP_000160.1; Met1-Leu429). alpha-Galactosidase A / GLA specific IgG was purified by Human alpha-Galactosidase A / GLA affinity chromatography.
Catalog No. 50-241-0210