Learn More
Biomatik Corporation Human Glial Cell Line Derived Neurotrophic Factor Receptor Alpha 1 (GFRa1) ELISA Kit
Supplier:  Biomatik Corporation EKU04378
Encompass_Preferred
Human Glial Cell Line Derived Neurotrophic Factor Receptor Alpha 1 (GFRa1) ELISA Kit for detecting GFRa1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P56159. Gene ID: 2674. Target Synonyms: GDNFR; GDNFRA; GFR-ALPHA-1; RET1L; RETL1; TRNR1; GDNF family receptor alpha-1; TGF-beta-related neurotrophic factor receptor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8959