Learn More
Biomatik Corporation Human Glial Cell Line Derived Neurotrophic Factor (GDNF) CLIA Kit
Supplier:  Biomatik Corporation EKU09437
Encompass_Preferred
Human Glial Cell Line Derived Neurotrophic Factor (GDNF) CLIA Kit for detecting GDNF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 4.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P39905. Gene ID: 2668. Target Synonyms: ATF1; ATF2; HFB1-GDNF; Astrocyte-Derived Trophic Factor; Glial Derived Neurotrophic Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8520