Learn More
Raybiotech Inc Human GLP-1 ELISA Kit
Supplier:  Raybiotech Inc ELHGLP736A1
Encompass_Preferred
For cell culture supernatants, EDTA treated plasma, and serum samples. Heparin and citrate are not recommended. Sensitivity: 150 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human GLP-1 (7-36a). Other species not determined
Catalog No. 50-194-9319