Learn More
Sino Biological Recombinant Human GLP1R Protein (VLP, aa24-463, Flag & Strep Tag)
Catalog No. 502993555
Encompass_Preferred
A DNA sequence encoding the Human GLP1R-VLP (P43220) (Arg24-Ser463) was expressed, with a Flag tag at the N-terminus, and a strep tag at the C-terminus.
Catalog No. 50-299-3555 Supplier Sino Biological Supplier No. 13944H84H20
A DNA sequence encoding the Human GLP1R-VLP (P43220) (Arg24-Ser463) was expressed, with a Flag tag at the N-terminus, and a strep tag at the C-terminus.