Learn More
Sino Biological Recombinant Human GLP1R Protein (Nanodisc, Flag-His Tag)
Catalog No. 502993273
Encompass_Preferred
A DNA sequence encoding the Human GLP1R (P43220) (Arg24-Phe257) & (Glu262-Lys437) was expressed and linked by a Fusion protein, with polyhistidine tag and Flag tag at the N-terminus. Within the expressed regions, there are several proprietary thermostabilizing mutations. This protein was prepared using our Nanodisc platform.Nanodisc is a versatile tool for studying membrane proteins. Using styrene-maleic acid (SMA) copolymer, membrane proteins can be extracted directly from prokaryotic and eukaryotic expression systems in the absence of detergents to preserve the protein structure and function better. Compared to membrane scaffold proteins (MSPs) nanodiscs, SMA nanodiscs also have the advantage of preserving proteins' nature by maintaining native lipids surrounded without introducing any heterologous proteins, which allows studies of protein structure and functions in a native-like environment.
Catalog No. 50-299-3273 Supplier Sino Biological Supplier No. 13944H88HNA20
A DNA sequence encoding the Human GLP1R (P43220) (Arg24-Phe257) & (Glu262-Lys437) was expressed and linked by a Fusion protein, with polyhistidine tag and Flag tag at the N-terminus. Within the expressed regions, there are several proprietary thermostabilizing mutations. This protein was prepared using our Nanodisc platform.Nanodisc is a versatile tool for studying membrane proteins. Using styrene-maleic acid (SMA) copolymer, membrane proteins can be extracted directly from prokaryotic and eukaryotic expression systems in the absence of detergents to preserve the protein structure and function better. Compared to membrane scaffold proteins (MSPs) nanodiscs, SMA nanodiscs also have the advantage of preserving proteins' nature by maintaining native lipids surrounded without introducing any heterologous proteins, which allows studies of protein structure and functions in a native-like environment.