Learn More
ACROBiosystems Human GLP1R Full-Length Protein (VLP), HEK293 Expressed, Arg 24-Ser 432, 100 μg
Catalog No. NC3878102
Encompass_Preferred
Recombinant full-length human GLP1R provided as virus-like particles (VLP), expressed in HEK293 cells and covering amino acids Arg 24-Ser 432. Supplied in a PBS-based formulation with arginine and trehalose; store at -70°C or lower. Intended for immunization and antibody screening.
  • Full-length human GLP1R, amino acids Arg 24-Ser 432.
  • Expressed in HEK293 (human 293) cells.
  • Supplied in virus-like particle (VLP) format.
  • Formulated in PBS with arginine, pH 7.4, and trehalose as protectant.
  • Store at -70°C or lower; stable for 12 months under sterile conditions.
  • Intended for immunization and antibody screening.
Catalog No. NC3878102 Supplier ACROBiosystems Supplier No. GLRH52P3100UG
Recombinant full-length human GLP1R provided as virus-like particles (VLP), expressed in HEK293 cells and covering amino acids Arg 24-Ser 432. Supplied in a PBS-based formulation with arginine and trehalose; store at -70°C or lower. Intended for immunization and antibody screening.
  • Full-length human GLP1R, amino acids Arg 24-Ser 432.
  • Expressed in HEK293 (human 293) cells.
  • Supplied in virus-like particle (VLP) format.
  • Formulated in PBS with arginine, pH 7.4, and trehalose as protectant.
  • Store at -70°C or lower; stable for 12 months under sterile conditions.
  • Intended for immunization and antibody screening.