Learn More
ACROBiosystems Human GLP1R Protein, Fc Tag
Supplier:  ACROBiosystems GLRH5253100UG
Encompass_Preferred
Human GLP1R Protein, Fc Tag (GLR-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Glu 139 (Accession # P43220-1).
Catalog No. 50-232-9109