Learn More
Biomatik Corporation Human Glucocorticoid Induced Tumor Necrosis Factor Receptor (GITR) ELISA Kit
Supplier:  Biomatik Corporation EKU04395
Encompass_Preferred
Human Glucocorticoid Induced Tumor Necrosis Factor Receptor (GITR) ELISA Kit for detecting GITR in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y5U5. Gene ID: 8784. Target Synonyms: TNFRSF18; AITR; Tumor Necrosis Factor Receptor Superfamily 18; Glucocorticoid-induced TNFR-related protein; Activation-inducible TNFR family receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4357