Learn More
Biomatik Corporation Human Glucocorticoid Receptor Alpha (GRa) ELISA Kit
Supplier:  Biomatik Corporation EKU04396
Encompass_Preferred
Human Glucocorticoid Receptor Alpha (GRa) ELISA Kit for detecting GRa in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04150. Gene ID: 2908. Target Synonyms: GR-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4462