Learn More
Invitrogen™ Human Glucocorticoid Receptor (aa 245-259) Synthetic Peptide
Description
PEP-002 is a 15 amino acid (amino acids 245-259) synthetic peptide whose sequence is from the amino terminus of the human glucocorticoid receptor (GR). The sequence of this peptide is (amino to carboxy terminus): C-K-P-L-I-L-P-D-T-K-P-K-I-K-D (The N-terminal cysteine has been added to facilitate conjugation and is not a part of the native human GR sequence) This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and the human GR, catalog # PA1-512. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody. Reconstitute with 0.1 mL of distilled water.
Specifications
Specifications
| Gene ID (Entrez) | 2908 |
| Species | Human |
| Purification Method | purified |
| Accession Number | P04150 |
| Common Name | Glucocorticoid Receptor (Nr3c1) |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | Peptide with no preservative |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.