Learn More
Biomatik Corporation Human Glucokinase (GCK) ELISA Kit
Supplier:  Biomatik Corporation EKU04400
Encompass_Preferred
Human Glucokinase (GCK) ELISA Kit for detecting GCK in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35557. Gene ID: 2645. Target Synonyms: GK; GLK; HHF3; HK4; HKIV; HXKP; MODY2; Hexokinase-D; Hexokinase 4; Maturity Onset Diabetes Of The Young 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5654