Learn More
Biomatik Corporation Human Glutamate Receptor Interacting Protein 1 (GRIP1) ELISA Kit
Supplier:  Biomatik Corporation EKU04445
Encompass_Preferred
Human Glutamate Receptor Interacting Protein 1 (GRIP1) ELISA Kit for detecting GRIP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y3R0. Gene ID: 23426. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5744