Learn More
Biomatik Corporation Human Glutamate Receptor, Ionotropic, AMPA 1 (GRIA1) ELISA Kit
Supplier:  Biomatik Corporation EKU04446
Encompass_Preferred
Human Glutamate Receptor, Ionotropic, AMPA 1 (GRIA1) ELISA Kit for detecting GRIA1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P42261. Gene ID: 2890. Target Synonyms: GLUH1; GLUR1; GLURA; HBGR1; GluR-K1; AMPA-selective glutamate receptor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8955