Learn More
Biomatik Corporation Human Glutamate Receptor, Ionotropic, Delta 2 (GRId2) ELISA Kit
Supplier:  Biomatik Corporation EKU09294
Encompass_Preferred
Human Glutamate Receptor, Ionotropic, Delta 2 (GRId2) ELISA Kit for detecting GRId2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O43424. Gene ID: 2895. Target Synonyms: GluD2; GLURD2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8540