Learn More
Biomatik Corporation Human Glutamate Receptor, Ionotropic, AMPA 3 (GRIA3) ELISA Kit
Supplier:  Biomatik Corporation EKU04450
Encompass_Preferred
Human Glutamate Receptor, Ionotropic, AMPA 3 (GRIA3) ELISA Kit for detecting GRIA3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P42263. Gene ID: 2892. Target Synonyms: GLUR-K3; GLUR3; GLURC; gluR-C; AMPA-selective glutamate receptor 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8954