Learn More
Biomatik Corporation Human Glutamate Receptor, Ionotropic, N-Methyl-D-Aspartate 2D (GRIN2D) ELISA Kit
Supplier:  Biomatik Corporation EKU04459
Encompass_Preferred
Human Glutamate Receptor, Ionotropic, N-Methyl-D-Aspartate 2D (GRIN2D) ELISA Kit for detecting GRIN2D in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.123ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O15399. Gene ID: 2906. Target Synonyms: EB11; NMDAR2D; NR2D; N-Methyl-d-Aspartate Receptor Subunit 2D; Glutamate [NMDA] receptor subunit epsilon-4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4974