Learn More
Biomatik Corporation Human Glutamate Receptor, Metabotropic 1 (GRM1) ELISA Kit
Supplier:  Biomatik Corporation EKU04460
Encompass_Preferred
Human Glutamate Receptor, Metabotropic 1 (GRM1) ELISA Kit for detecting GRM1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13255. Gene ID: 2911. Target Synonyms: GPRC1A; GRM1A; MGLUR1; MGLUR1A; mGluR1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6164