Learn More
American Research Products Inc Human glutamic acid decarboXyl
Catalog No. 5000329312
Encompass
Human glutamic acid decarboXylase 65 GAD65 ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29312 Supplier American Research Products Inc Supplier No. CSBE14119H
Human glutamic acid decarboXylase 65 GAD65 ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich