Learn More
Biomatik Corporation Human Glutamine Fructose-6-Phosphate Transaminase 2 (GFPT2) ELISA Kit
Supplier:  Biomatik Corporation EKU04465
Encompass_Preferred
Human Glutamine Fructose-6-Phosphate Transaminase 2 (GFPT2) ELISA Kit for detecting GFPT2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O94808. Gene ID: 9945. Target Synonyms: GFAT2; Glutamine Fructose 6-Phosphate Aminotransferase 2; Hexosephosphate aminotransferase 2; D-fructose-6-phosphate amidotransferase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5810