Learn More
Abcam Human Glutamine Synthase ELISA Kit, 1 x 96 Tests
Catalog No. 502654628
Encompass_Preferred
Human Glutamine Synthase ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Glutamine Synthase with a sensitivity of 18 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-4628 Supplier Abcam Supplier No. AB2889271X96TESTS
Human Glutamine Synthase ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Glutamine Synthase with a sensitivity of 18 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement